Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Vermiform Appendix Section
SCI_ED

Catalog No. S30386 Shop All Eisco Products
Change view
Click to view available options
Grade:
9 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S30386 9 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30386 Supplier Eisco™ Supplier No. BS18245

Prepared microscope slide of a vermiform appendix. Showing characteristic epithelial layers of digestive system. Great choice for biology classrooms to explore structure function relationships as per NGSS standards.

  • pre-cleaned glass slide with ground edges
  • size 1x3 in. (25x77 mm)
  • professionally stained samples for best visualization
  • great choice for biology classrooms to explore structure-function relationships as per NGSS standards
  • showing characteristic epithelial layers of digestive system

Specifications

Description Shows characteristic epithelial layer
Classification Digestive System
Color Clear
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Grade 9 to 12
Quantity 1
Specimen Type Vermiform Appendix
Size 1 x 3 in.
Slide Type Prepared Slide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.