Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Smooth Muscle (Mammal), Cross Section
SCI_ED

Catalog No. S30364 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S30364 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30364 Supplier Eisco™ Supplier No. BS18219
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Prepared microscope slide of a cross section of mammalian smooth muscle. Showing characteristic morphology and structures of smooth muscle cells. Great choice for biology classrooms to explore structure function relationships as per NGSS standards.

  • Pre-cleaned glass slide with ground edges
  • Size 1x3 in. (25x77 mm)
  • Professionally stained samples for best visualization
  • Great choice for biology classrooms to explore structure-function relationships as per NGSSs standards
  • Showing characteristic morphology and structures of smooth muscle cells

Specifications

Description Cross Section shows characteristic morphology and structures of smooth muscle cells
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Quantity 1 Ea.
Species Mammal
Specimen Type Smooth Muscle
Size 1 x 3 in.
Slide Type Prepared Slide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.