Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Kidney Section (Human), Histological Structures
SCI_ED

Catalog No. S30365 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S30365 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30365 Supplier Eisco™ Supplier No. BS18220
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Prepared microscope slide of a section of human kidney. Showing characteristic histological structures. Great choice for biology classrooms to explore structure function relationships as per NGSS standards.

  • Pre-cleaned glass slide with ground edges
  • Size 1x3 in. (25x77 mm)
  • Professionally stained samples for best visualization
  • Great choice for biology classrooms to explore structure-function relationships as per NGSSs standards
  • Showing characteristic histological structures

Specifications

Classification Histology
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Quantity 1 Ea.
Species Human
Specimen Type Kidney Section
Size 1 x 3 in.
Slide Type Prepared Slide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.