Learn More
Eisco™ Apple Tree Stem, Cross Section
SCI_ED
Catalog No. S30654 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
S30654 1 Ea.
1 options
Catalog No. S30654 Supplier Eisco™ Supplier No. BS18298
Add to Cart
Add to Cart
Prepared microscope slide of a cross section of apple tree stem. Apple is a dicot. Stained for better visualization of characteristic dicot stem structures. Great choice for biology classrooms to explore structure function relationships as per NGSS standards.
  • Pre-cleaned glass slide with ground edges
  • Size 1x3 in. (25x77 mm)
  • Professionally stained samples for best visualization
  • Great choice for biology classrooms to explore structure-function relationships as per NGSSs standards
  • Showing characteristic dicot stem structures
Specifications
Description Cross section of an Apple Tree stem, stained for better visualization of characteristic dicot stem structures
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Quantity 1 Ea.
Specimen Type Apple Tree Stem
Size 1 x 3 in.
Slide Type Prepared Slide