Learn More
Eisco™ Lilium Anther, Cross Section
SCI_ED
Catalog No. S30362 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
S30362 1 Ea.
1 options
Catalog No. S30362 Supplier Eisco™ Supplier No. BS18217
Add to Cart
Add to Cart
Prepared microscope slide of a cross section of a mature lillium anther containing pollen. Anther is a part of the flower where pollen is produced. Showing characteristic structures of lilium anther and pollen. Great choice for biology classrooms.
  • Pre-cleaned glass slide with ground edges
  • Size 1x3 in. (25x77 mm)
  • Professionally stained samples for best visualization
  • Great choice for biology classrooms to explore structure-function relationships as per NGSSs standards
  • Showing characteristic structures of lilium anther and pollen
Specifications
Description Cross section of a mature Lillium anther containing pollen, shows characteristic structures
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Quantity 1 Ea.
Specimen Type Lilium Anther
Size 1 x 3 in.
Slide Type Prepared Slide