Learn More
Eisco™ Lilium Ovary
SCI_ED
Catalog No. S30673 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
S30673 1 Ea.
1 options
Catalog No. S30673 Supplier Eisco™ Supplier No. BS18322
Add to Cart
Add to Cart
Prepared microscope slide of lilium ovary. Stained for better visualization of characteristic structures including ovules and embryo sacs. Great choice for biology classrooms to explore structure function relationships as per NGSS standards.
  • Pre-cleaned glass slide with ground edges
  • Size 1x3 in. (25x77 mm)
  • Professionally stained samples for best visualization
  • Great choice for biology classrooms to explore structure-function relationships as per NGSSs standards
  • Showing characteristic structures
Specifications
Description Lilium Ovary, stained for better visualization of ovules and embryo sacs
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Quantity 1 Ea.
Specimen Type Lilium Ovary
Size 1 x 3 in.
Slide Type Prepared Slide