Learn More
Eisco™ Moss Archegonia, Longitudinal Section
SCI_ED
Catalog No. S30352 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
S30352 1 Ea.
1 options
Catalog No. S30352 Supplier Eisco™ Supplier No. BS18205
Add to Cart
Add to Cart
Prepared microscope slide of a lontitudinal section of moss archegonia. Archegonia are organs which produce and contain ova or female gametes. Great choice for biology classrooms studying sexual reproduction of plants and fungi.
  • Pre-cleaned glass slide with ground edges
  • Size 1x3 in. (25x77 mm)
  • Professionally stained samples for best visualization
  • Great choice for biology classrooms to explore structure-function relationships as per NGSSs standards
  • Showing characteristic structures including females gametes
Specifications
Description Longtitudinal section of Moss Archegonia, organs that produce and contain ova or female gametes
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Quantity 1 Ea.
Specimen Type Moss Archegonia
Size 1 x 3 in.
Slide Type Prepared Slide