Learn More
Eisco™ Tilia 1 Yr Stem, Cross Section
SCI_ED
Catalog No. S30381 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1
Catalog No. Quantity
S30381 1
1 options
Catalog No. S30381 Supplier Eisco™ Supplier No. BS18239
Add to Cart
Add to Cart
Prepared microscope slide of one year old tilia stem cross section. Tilia, commonly known as basswood is a dicot. Showing characteristic structures of a dicot stem. Great choice for biology classrooms to explore structure function relationships as per NGSS standards.
  • pre-cleaned glass slide with ground edges
  • size 1x3 in. (25x77 mm)
  • professionally stained samples for best visualization
  • great choice for biology classrooms to explore structure-function relationships as per NGSS standards
  • showing characteristic structures of a dicot stem
Specifications
Description Shows characteristic structures of a one year old dicot stem.
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Quantity 1
Specimen Type Tilia Stem
Size 1 x 3 in.
Slide Type Prepared Slide