Learn More
Eisco™ Onion Root Tip Squash, Whole Mount
SCI_ED
Catalog No. S30348 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
S30348 1 Ea.
1 options
Catalog No. S30348 Supplier Eisco™ Supplier No. BS18201
Add to Cart
Add to Cart
Prepared microscope slide of an onion root tip squash. Whole mount. Specially stained to show cells in all stages of mitotic division, as well as other characteristic structures of a plant cell. Great tool for biology classrooms studying cell division
  • Pre-cleaned glass slide with ground edges
  • Size 1x3 in. (25x77 mm)
  • Professionally stained samples for best visualization
  • Great choice for biology classrooms to explore structure-function relationships as per NGSSs standards
  • Showing cells in all stages of mitotic division, as well as other characteristic structures of a plant cell
Specifications
Description Whole mount is stained to specifically show cells in all stages of mitotic division
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Quantity 1 Ea.
Specimen Type Onion Root Tip Squash
Size 1 x 3 in.
Slide Type Prepared Slide