Learn More
Eisco™ Tilia Two and Three Year Old Stem (Basswood), Cross Section
SCI_ED
Catalog No. S30360 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
S30360 1 Ea.
1 options
Catalog No. S30360 Supplier Eisco™ Supplier No. BS18215
Add to Cart
Add to Cart
Prepared microscope slide of a cross section of a stem of tillia, commonly known as basswood. This slide shows a composite section of a two and three year old basswood stem, for better visualization of structural changes in dicot stem over time.
  • Pre-cleaned glass slide with ground edges
  • Size 1x3 in. (25x77 mm)
  • Professionally stained samples for best visualization
  • Great choice for biology classrooms to explore structure-function relationships as per NGSSs standards
  • Showing characteristic structures of two and three year old tilia stem, side by side for easy observation of its development over time
Specifications
Description Cross section of a two and three year old Tilia depicts structural changes in a dicot stem over time
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Quantity 1 Ea.
Specimen Type Tilia (Basswood)
Size 1 x 3 in.
Slide Type Prepared Slide