Learn More
Description
- Pre-cleaned glass slide with ground edges
- Size 1x3 in. (25x77 mm)
- Professionally stained samples for best visualization
- Great choice for biology classrooms to explore structure-function relationships as per NGSSs standards
- Showing characteristic root tip structures
Specifications
Specifications
| Description | Wholemount Wheat Root Tip shows characteristic structures |
| Classification | Botany |
| Material | Glass |
| Type | Prepared Microscope Slide |
| For Use With (Application) | Microscopy |
| Quantity | 1 Ea. |
| Specimen Type | Wheat Root Tip |
| Size | 1 x 3 in. |
| Slide Type | Prepared Slide |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
