Learn More
Eisco™ Lilium Ovary, Cross Section
SCI_ED
Catalog No. S30356 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
S30356 1 Ea.
1 options
Catalog No. S30356 Supplier Eisco™ Supplier No. BS18210
Add to Cart
Add to Cart
Prepared microscope slide of a cross section of lilium ovary. Stained for better visualization of characteristic structures including placenta, ovule, and locule. Great choice for biology classrooms to explore structure function relationships as per NGSS standards.
  • Pre-cleaned glass slide with ground edges
  • Size 1x3 in. (25x77 mm)
  • Professionally stained samples for best visualization
  • Great choice for biology classrooms to explore structure-function relationships as per NGSSs standards
  • Showing characteristic structures including placenta, ovule, and locule
Specifications
Description Cross section of Lilum Ovary shows characteristic placenta, ovule, and locule
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Quantity 1 Ea.
Specimen Type Lilium Ovary - Cross Section
Size 1 x 3 in.
Slide Type Prepared Slide