Learn More
Eisco™ Elodea Stem Tip, Cross Section
SCI_ED
Catalog No. S30651 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
S30651 1 Ea.
1 options
Catalog No. S30651 Supplier Eisco™ Supplier No. BS18295
Add to Cart
Add to Cart
Prepared microscope slide of a cross section of eleodea stem tip. Stem tip contains apical meristem and is a location of plant growth. Stained for better visualization of characteristic structures. Great choice for biology classrooms to explore structure function relationships as per NGSS standards.
  • Pre-cleaned glass slide with ground edges
  • Size 1x3 in. (25x77 mm)
  • Professionally stained samples for best visualization
  • Great choice for biology classrooms to explore structure-function relationships as per NGSSs standards
  • Showing characteristic structures and tissues
Specifications
Description Cross section of Eleodea stem tip that contains apical meristem, a location of plant growth
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Quantity 1 Ea.
Specimen Type Elodea Stem Tip
Size 1 x 3 in.
Slide Type Prepared Slide