Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Fern Prothallium, Cross Section
SCI_ED

Catalog No. S30668 Shop All Eisco Products
Change view
Click to view available options
Grade:
9 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S30668 9 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30668 Supplier Eisco™ Supplier No. BS18316

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

Prepared microscope slide of a cross section of fern prothallium. Prothallium is a gametophyte stage in the life cycle of fern. Showing general structures of Prothallium. A great tool for studying fern life cycle and reproduction.

  • Pre-cleaned glass slide with ground edges
  • Size 1x3 in. (25x77 mm)
  • Professionally stained samples for best visualization
  • Great choice for biology classrooms to explore structure-function relationships as per NGSSs standards
  • Showing general structures of Prothallium

Specifications

Description Cross section of Fern Prothallium, a gametophyte stage in the life cycle of fern
Classification Botany
Color Clear
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Grade 9 to 12
Quantity 1 Ea.
Specimen Type Fern Prothallium
Size 1 x 3 in.
Slide Type Prepared Slide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.