Learn More
Eisco™ Salix Branch Root Origin, Cross Section
SCI_ED
Catalog No. S30371 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1
Catalog No. Quantity
S30371 1
1 options
Catalog No. S30371 Supplier Eisco™ Supplier No. BS18226
Add to Cart
Add to Cart
Prepared microscope slide of a section of salix branch root origin. Stained for easy visualization of the origin of the root in a new branch cutting of a willow. Showing characteristic structures. Great choice for biology classrooms.
  • pre-cleaned glass slide with ground edges
  • size 1x3 in. (25x77 mm)
  • professionally stained samples for best visualization
  • great choice for biology classrooms to explore structure-function relationships as per NGSS standards
  • showing origin of the root in a new branch cutting of a willow
Specifications
Description Easy visualization of the root origin in a new branch cutting of a willow
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Quantity 1
Specimen Type Salix Branch Root Origin
Size 1 x 3 in.
Slide Type Prepared Slide