Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Monocot/Dicot Root, Composite
SCI_ED

Catalog No. S30374 Shop All Eisco Products
Change view
Click to view available options
Grade:
9 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S30374 9 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30374 Supplier Eisco™ Supplier No. BS18229
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Prepared composite microscope slide of monocot and dicot root. Showing side by side monocot and dicot root for easy comparison of characteristic monocot and dicot root structures. Great choice for biology classrooms to explore structure function relationships as per NGSS standards.

  • pre-cleaned glass slide with ground edges
  • size 1x3 in. (25x77 mm)
  • professionally stained samples for best visualization
  • great choice for biology classrooms to explore structure-function relationships as per NGSS standards
  • showing side by side monocot and dicot root for easy comparison of characteristic monocot and dicot root structures

Specifications

Description Side by side monocot and dicot root for easy comparison of characteristic root structures
Classification Botany
Color Clear
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Grade 9 to 12
Quantity 1
Specimen Type Monocot and Dicot Root
Size 1 x 3 in.
Slide Type Prepared Slide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.