Learn More
BD Prepared Bottled Media: Tryptic Soy Broth, Wide Mouth Bottle
Shop All BD Diagnostic Systems ProductsDescription
- Ready for immediate use
- Sterility Assurance Level of 10-6 reduces risk of false positives
- Terminally autoclaved
- Conforms to Harmonized USP, EP, and JP requirements for sterility tests and to CLSI requirements for recovery
Specifications
Specifications
| Cap Type | Lug cap |
| Color | Amber |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | Tryptic Soy Broth |
| Product Type | Broth |
| Fill Volume | 100 mL |
| Packaging Type | Bottle |
| Quantity | 100 mL |
| Height (Metric) | 130 mm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.