Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prepared Culture Media in Bottles
SCI_ED

Catalog No. s716944a Shop All Fischer Technical Company Products
Change view
Click to view available options
Media Type:
Nutrient Agar
Nutrient Broth
Plate Count Agar
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Media Type
S716944A Nutrient Broth
S716934A Nutrient Agar
S716954A Plate Count Agar
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S716944A Supplier Fischer Technical Company Supplier No. CM146B125
Add to Cart
Add to Cart

Media stay fresh in these disposable bottles.

  • Each bottle is labeled with product name, expiration date and lot number for easy handling
  • Bottles are narrow-mouth with black phenolic screw caps
  • Capacity: 125mL each

Order Info

Sold in packs of 24 bottles.

Specifications

Form Liquid
Manufacturing Origin United States
Media Type Nutrient Broth
Product Type Prepared Microbiology Media
Sterility Sterile
Target Organism Class Bacteria
Test Type Microbial Identification
Fill Volume 125 mL
Packaging Type Bottle
Quantity 125 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.