Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fischer Technical Company™ Prepared Culture Media in Plates
SCI_ED

Catalog No. S012399 Shop All Fischer Technical Company Products
Change view
Click to view available options
Media Type:
LB Agar
MacConkey Agar
Nutrient Agar
Plate Count Agar
Potato Dextrose Agar
Sabouraud Dextrose Agar
Tryptic Soy Agar
Detects:
Bacteria
Fungi
Gram Negative Bacteria
Microbes
Non-fastidious Aerobic and Anaerobic Microorganisms
Non-fastidious Microorganisms
Yeast, Mold
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Media Type Detects
S012399 MacConkey Agar Gram Negative Bacteria
S716961A Potato Dextrose Agar Yeast, Mold
S716951A Plate Count Agar Microbes
S716931A Nutrient Agar Non-fastidious Microorganisms
S716981A Tryptic Soy Agar Non-fastidious Aerobic and Anaerobic Microorganisms
S716971A Sabouraud Dextrose Agar Fungi
S012390 LB Agar Bacteria
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. S012399 Supplier Fischer Technical Company™ Supplier No. CM102P25
Add to Cart
Add to Cart

Prepared Culture Media in Plates labeled with product name, expiration date and lot number.

  • Sterile plates are ready for immediate use
  • Supplied in disposable plastic petri dishes
  • Packed in clear, heat-sealed cellophane bags

Specifications

Detects Gram Negative Bacteria
Format Plate
Manufacturing Origin US with Globally Sourced Materials
Media Type MacConkey Agar
Product Type Agar Plate
Sterility Sterile
Target Organism Class Gram-negative Bacteria
Test Type Microbiological Tests
For Use With (Application) Microbiological Testing
Quantity 10 Plates
Height (English) 0.59 in.
Height (Metric) 15 mm
Diameter (English) Outside 3.93 in.
Diameter (Metric) Outside 100 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.