Learn More
Fischer Technical Company™ Prepared Culture Media in Plates
Shop All Fischer Technical Company Products

Description
- Sterile plates are ready for immediate use
- Supplied in disposable plastic petri dishes
- Packed in clear, heat-sealed cellophane bags
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Detects | Non-fastidious Aerobic and Anaerobic Microorganisms |
| Format | Plate |
| Manufacturing Origin | US with Globally Sourced Materials |
| Media Type | Tryptic Soy Agar |
| Product Type | Prepared Microbiology Plate |
| Sterility | Sterile |
| Target Organism Class | Aerobic Microorganisms |
| Test Type | Microbial Identification |
| For Use With (Application) | Microbiological Testing |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.