Learn More
Fischer Technical Company™ Prepared Culture Media in Tubes
Shop All Fischer Technical Company Products

Description
- Tubes measure 16 mm O.D. x 125 mm long
- Include black plastic screw caps
- Each tube is labeled with product name, expiration date and lot number
- Refrigerate until ready to use
Order Info
Sold in pack of 20 tubes.
Specifications
Specifications
| Manufacturing Origin | US with Globally Sourced Materials |
| Media Type | Nutrient Broth |
| Product Type | Prepared Microbiology Tubes |
| Sterility | Sterile |
| Target Organism Class | Bacteria |
| Test Type | Microbial Identification |
| Fill Volume | 10 mL |
| Quantity | 20 Tubes/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.