Learn More
Prepared Individual Microscope Slides
SCI_ED
Catalog No. s99469 Shop All Johannes Lieder GmbH & Co. KG Products
Change view
Click to view available options
Specimen Type:
Beef Liver Fluke
Blood
Head and Mouth Parts w/ Sucking Tube
Liver
Older Stem w/ Annual Rings, Resin Ducts
Radiolaria, Mixed Species
Spirogyra
Typical Dicot Herbaceous Stem
Species:
Fasciola hepatica
Helianthus, Sunflower
Human
Musca domestica, House Fly
Pig
Pinus
Radiolaria, Mixed Species
Spirogyra
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Specimen Type Species
S99469 Older Stem w/ Annual Rings, Resin Ducts Pinus
S99461 Blood Human
S99455 Radiolaria, Mixed Species Radiolaria, Mixed Species
S99472 Typical Dicot Herbaceous Stem Helianthus, Sunflower
S99468 Spirogyra Spirogyra
S99458 Head and Mouth Parts w/ Sucking Tube Musca domestica, House Fly
S99463 Liver Pig
S99456 Beef Liver Fluke Fasciola hepatica
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. S99469 Supplier Johannes Lieder GmbH & Co. KG Supplier No. GY125C
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
High-quality microscope slides, manufactured under rigorous control to meet all scientific requirements.
  • Premium glass with fine ground edges
  • Size: 1 x 3 in. (26 x 76mm)
  • Specially stained to ensure a clear, multicolored representation of all tissue structures
  • Specimens are characteristic and representative of the taxonomic group and/or subject
  • All slides sold separately
Specifications
Material Glass
Mounting Type Cross and Longitudinal Section
Type Pinus, older stem with annual rings, resin ducts
Species Pinus
Specimen Type Older Stem w/ Annual Rings, Resin Ducts
Size 1 x 3 in.
Slide Type Prepared Slide