Learn More
Eisco™ Bacteria Spores Smear
SCI_ED
Catalog No. S30366 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
S30366 1 Ea.
1 options
Catalog No. S30366 Supplier Eisco™ Supplier No. BS18221
Add to Cart
Add to Cart
Prepared microscope slide of a smear of bacterial spores. Bacterial spores are highly resistant dormant structures that bacteria form in response to adverse environmental conditions. Showing characteristic morphology.
  • Pre-cleaned glass slide with ground edges
  • Size 1x3 in. (25x77 mm)
  • Professionally stained samples for best visualization
  • Great choice for biology classrooms to explore structure-function relationships as per NGSSs standards
  • Showing characteristic morphology
Specifications
Description Bacterial Spores are highly resistant, dormant structures that are formed in response to adverse environmental conditions
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Quantity 1 Ea.
Specimen Type Bacteria Spores
Size 1 x 3 in.
Slide Type Prepared Slide