Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Prepared Microscope Slide - Ground Compact Bone
SCI_ED

Catalog No. s29632 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S29632 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S29632 Supplier Eisco™ Supplier No. BS18017
Only null left
Add to Cart
Add to Cart

Prepared microscope slide of compact bone

  • Pre-cleaned glass slide with ground edges
  • Professionally stained samples for best visualization
  • Ideal for biology classrooms to explore structure-function relationships as per NGSS standards
  • Shows general structures of calcified bone tissue
  • Ground bone is prepared without de-calcifying the bone to make a thin section

Specifications

Biosafety Level 1
Concentration 10%
Material Glass
Type Microbiology Slide
For Use With (Application) Microscopy
Manufacturing Origin China
Media Type Formalin
Packaging Cardboard Mailer
Quantity 1
Species Human or Pig
Size 1 x 3 in.
Slide Type Prepared Slide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.