Learn More
United Scientific™ Prepared Microscope Slide Set of 14
SDPSCI_ED
Catalog No. S111013 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Slide Type:
Prepared Slide
Catalog No. Slide Type
S111013 Prepared Slide
1 options
Catalog No. S111013 Supplier United Scientific™ Supplier No. SLD014
Add to Cart
Add to Cart
This Prepared Microscope Slide Set features a convenient assortment of prepared microscope slides that includes 14 samples affixed to quality glass. Case included.
Set includes:
  • Allium Mitosis, ls
  • Bone, Ground, cs
  • Desmids, wm
  • Epithelium, Squamous, sm
  • Ficus Leaf, cs
  • Giant Multipolar Motor Neurons
  • Muscle Types Composite, sec
  • Ranunculus - Stem, cs
  • Skin, Cornified, sec
  • Spirogyra - Vegetative, wm
  • Tilia - Meristematic Stem, cs
  • Trachea, Mammal, ls
  • Volvox, sec
  • Zea (Corn) Stem, cs
Specifications
Material Glass
Type Prepared Microscope Slide Set
For Use With (Application) Used under Microscope
Includes Plastic Case
Manufacturing Origin China
Packaging Set of 14
Quantity 1 Ea.
Slide Type Prepared Slide