Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Prepared Wash Buffer (WBC) for ReliaPrep™ Large Volume HT gDNA Isolation System

Catalog No. PRA2681 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
3.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRA2681 3.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRA2681 Supplier Promega Supplier No. A2681
Add to Cart
Add to Cart

Prepared Wash Buffer (WBC) is a component of the ReliaPrep Large Volume HT gDNA Isolation System (Cat.# A2751) that is available for purchase separately.

  • Standalone Buffer Component of the ReliaPrep™ Large Volume HT gDNA Isolation System
  • Used for automated DNA purification from large-volume blood samples with the ReliaPrep™ Large Volume HT gDNA Isolation System on liquid handlers

Specifications

Content And Storage 15°C to 30°C
Quantity 3.5 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.