Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Corpus Luteum Section (Mammal)
SCI_ED

Catalog No. S30624 Shop All Eisco Products
Change view
Click to view available options
Grade:
9 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S30624 9 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30624 Supplier Eisco™ Supplier No. BS18275

Prepared microscope slide of mammalian corpus luteum. Corpus luteum is a temporary endocrinne gland that is created in the ovary after ovulation. Great choice for biology classrooms to explore structure function relationships as per NGSS standards.

  • Pre-cleaned glass slide with ground edges
  • Size 1 x 3 in. (25 x 77 mm)
  • Professionally stained samples for best visualization
  • Great choice for biology classrooms to explore structure-function relationships per NGSSs standards
  • Showing corpus luteum as well as developing follicles

Specifications

Description Mammalian Corpus Luteum, a temporary endocrinne gland created in the ovary after ovulation
Classification Animal Development
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Grade 9 to 12
Quantity 1 Ea.
Specimen Type Corpus Luteum
Size 1 x 3 in.
Slide Type Prepared Slide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.