Learn More
Eisco™ Frog Intestine, Cross Sectin and H&E Stain
SCI_ED
Catalog No. S30657 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
S30657 1 Ea.
1 options
Catalog No. S30657 Supplier Eisco™ Supplier No. BS18301
Add to Cart
Add to Cart
Prepared microscope slide of a cross section of frog intestine. Stained with hematoxylin and eosin for better visualization of characteristic structures and tissue layers. Great choice for biology classrooms to explore structure function relationships as per NGSS standards.
  • Pre-cleaned glass slide with ground edges
  • Size 1x3 in. (25x77 mm)
  • Professionally stained samples for best visualization
  • Great choice for biology classrooms to explore structure-function relationships as per NGSSs standards
  • Showing characteristic structures and tissue layers
Specifications
Description Cross section of Frog Intestine stained for better visualization of characteristic structures and tissue layers
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Quantity 1 Ea.
Specimen Type Frog Intestine
Stain Type H and E stain
Size 1 x 3 in.
Slide Type Prepared Slide