Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Frog Skin Section
SCI_ED

Catalog No. S30594 Shop All Eisco Products
Change view
Click to view available options
Grade:
9 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S30594 9 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30594 Supplier Eisco™ Supplier No. BS18252
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Prepared microscope slide of a section of frog skin. Showing the muscle, dermis and epidermis layers as well as characteristic structures such as mucus glands, blood vessels and chormatophores. Great choice for biology classrooms.

  • Pre-cleaned glass slide with ground edges
  • Size 1 x 3 in. (25 x 77 mm)
  • Professionally stained samples for best visualization
  • Great choice for biology classrooms to explore structure-function relationships per NGSSs standards
  • Showing muscle, dermis and epidermis layers as well as characteristic structures such as mucus glands, blood vessels and chormatophores

Specifications

Description Frog Skin section showing the muscle, dermis, and epidermis layers. Also shows characteristic structures such as the mucus glands, blood vessels, and chormatophores.
Classification Zoology
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Grade 9 to 12
Quantity 1 Ea.
Species Frog
Specimen Type Frog Skin
Size 1 x 3 in.
Slide Type Prepared Slide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.