Learn More
Eisco™ Mixed Rotifers (Fresh Water), Whole Mount
SCI_ED
Catalog No. S30618 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
S30618 1 Ea.
1 options
Catalog No. S30618 Supplier Eisco™ Supplier No. BS18269
Add to Cart
Add to Cart
Prepared microscope slide of wholemount of mixed freshwater rotifers. Showing characteristic morphology and structures. Great choice for biology classrooms to explore structure function relationships as per NGSS standards.
  • Pre-cleaned glass slide with ground edges
  • Size 1x3 in. (25x77 mm)
  • Professionally stained samples for best visualization
  • Great choice for biology classrooms to explore structure-function relationships as per NGSSs standards
  • Showing characteristic morphology and structures
Specifications
Description Wholemount of Mixed Freshwater Rotifers showing characteristic morphology and structures
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Quantity 1 Ea.
Specimen Type Mixed Fresh Water Rotifers
Size 1 x 3 in.
Slide Type Prepared Slide