Learn More
Eisco™ Uterus Section (Animal)
SCI_ED
Catalog No. S30620 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
S30620 1 Ea.
1 options
Catalog No. S30620 Supplier Eisco™ Supplier No. BS18271
Add to Cart
Add to Cart
Prepared microscope slide of animal uterus section. Uterus is a part of reproductive system. Stained for better visualization of characteristic layers and structures. Great choice for biology classrooms to explore structure function relationships as per NGSS standards.
  • Pre-cleaned glass slide with ground edges
  • Size 1x3 in. (25x77 mm)
  • Professionally stained samples for best visualization
  • Great choice for biology classrooms to explore structure-function relationships as per NGSSs standards
  • Showing characteristic layers and structures
Specifications
Description Animal Uterus section stained for better visualization of characteristic layers and structures
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Quantity 1 Ea.
Specimen Type Animal Uterus
Size 1 x 3 in.
Slide Type Prepared Slide