Description
- Pre-cleaned glass slide with ground edges
- Size 1x3 in. (25x77 mm)
- Professionally stained samples for best visualization
- Great choice for biology classrooms to explore structure-function relationships as per NGSSs standards
- Showing characteristic structures and morphology of ma mmalian kidney tissue
Specifications
Specifications
| Description | Cat Kidney stained for better visualization of mammalian kidney tissue |
| Material | Glass |
| Type | Prepared Microscope Slide |
| For Use With (Application) | Microscopy |
| Quantity | 1 Ea. |
| Specimen Type | Cat Kidney |
| Size | 1 x 3 in. |
| Slide Type | Prepared Slide |