Learn More
Eisco™ Reptile Blood Smear, GS Stain
SCI_ED
Catalog No. S30385 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1
Catalog No. Quantity
S30385 1
1 options
Catalog No. S30385 Supplier Eisco™ Supplier No. BS18244
Add to Cart
Add to Cart
Prepared microscope slide of reptile blood smear. Stained with Giemsa stain. Showing characteristic reptile blood cells and their relative ratios. Great choice for biology classrooms.
  • pre-cleaned glass slide with ground edges
  • size 1x3 in. (25x77 mm)
  • professionally stained samples for best visualization
  • great choice for biology classrooms to explore structure-function relationships as per NGSS standards
  • showing characteristic reptile blood cells and their relative ratios
Specifications
Description Shows characteristic reptile blood cells and their relative ratios
Material Glass
Type Prepared Microscope Slide
For Use With (Application) Microscopy
Quantity 1
Specimen Type Reptile Blood Smear
Stain Type Giemsa
Size 1 x 3 in.
Slide Type Prepared Slide