Learn More
Genscript Corporation 3C protease, His, Human
Supplier:  Genscript Corporation Z03092250
Encompass_Preferred
Recombinant PreScission Protease (rPSP) from human rhinovirus (HRV 3C) is a recombinant restriction-grade protease. PSP is a highly purified recombinant 6xHis-fusion protein, which recognizes the same cleavage site as the native enzyme. It is used to cleave affinity tags from fusion proteins. The optimum recognition site for this enzyme is the sequence Leu-Glu-Val-Leu-Phe-Gln/Gly-Pro(LEVLFQ/GP) and cleavage occurs between the Gln and Gly-Pro residues. The optimal temperature for cleavage is 4°C. It is recommended that the cleavage for each fusion protein be optimized by varying the amount of Recombinant PreScission Protease, reaction time, or incubation temperature. It can be removed by Ni2+ affinity resin.
Catalog No. NC1959008
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.