Learn More
Description
MA1-752 detects presenilin 1 protein (PS1) from mouse, rat, human, and nonhuman primate samples. No cross-reactivity is seen with presenilin 2. MA1-752 has successfully been used in immuno-fluorescence, immunocytochemistry, Western blot, immunoprecipitation, and ELISA procedures. By Western blot, this antibody detects an ~18 kDa protein representing PS1 CT (C-terminus fragment) and the ~46 kDa full-length PS1 from transfected SH-SY5Y cells. In 4% paraformaldehyde fixed N2a cells, MA1-752 showed endogenous PS1 staining in the Golgi and cellular vesicles. The MA1-752 immunogen is a synthetic peptide corresponding to residues C S(313) K Y N A E S T E R E S Q D T V A E N D D G(334) of humen PS1.
Specifications
Specifications
| Antigen | Presenilin 1 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | APS 18 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA and 0.05% sodium azide |
| Gene | PSEN1 |
| Gene Accession No. | P49768, P49769, P97887 |
| Gene Alias | AD3; Ad3h; Alzheimer Disease 3; FAD; Minilin; presenilin 1; presenilin 1 (Alzheimer disease 3); presenilin1; presenilin-1; Presenilin-1 CTF subunit; Presenilin-1 CTF12; Presenilin-1 NTF subunit; Protein S182; PS1; PS-1; PS1-CTF12; PSEN1; Psnl1; S182; S182 protein; Senilin 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
