Learn More
Poltex Pressure Blotter
SDP
Catalog No. 22000381
Change view
Click to view available options
Dimensions (L x W):
12 x 12 in.
Catalog No. Dimensions (L x W)
22-000-381 12 x 12 in.
1 options
Catalog No. 22-000-381 Supplier Poltex Supplier No. POSIBLOT1212
Add to Cart
Add to Cart
Replaces old Stratagene posiblot apparatus for Southern Blot applications.
Southern Blot positive pressure chamber.
Specifications
For Use With (Equipment) Stratagene pressure station
Includes Blotter chamber, membrane, gaskets
Material Acrylic
Dimensions (L x W) 12 x 12 in.
Hose and pressure sending unit are not included. Apparatus and gasket only.