Learn More
Grainger WATTS Pressure Regulator, 1/4INX1/4IN
Supplier:  Grainger 26X148
Encompass_Preferred
The Watts 560-26X148 is a 1/2" NPT bronze body pressure regulator with adjustable pressure settings from 25-75 psi. It has a maximum inlet pressure of 150 psi and a maximum temperature rating of 250°F. This regulator uses a diaphragm and spring to control downstream pressure and has a pressure relief valve for safety. It can be used on air, water, oil, and inert gas applications. The bronze body and trim provide corrosion resistance.
Catalog No. NC2383795
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.