Learn More
Selleck Chemical LLC Pyrroloquinoline Quinone Disodium Salt S5263-5mg
Supplier: Selleck Chemical LLC S52635MG
Pyrroloquinoline quinone (methoxatin disodium salt) an aromatic tricyclic o-quinone is a redox cofactor for bacterial dehydrogenases It is an efficient electron transfer catalyst from a number of organic substrates to molecular oxygen (O2) constructing quinoprotein model reactions
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.