Learn More
Thermo Scientific™ Pretreatment Cartridges and Filters
Catalog No. 10451115
Change view
Click to view available options
Type:
Filter
Filter with Stabilizer
Pretreatment Cartridge
Pretreatment Filter
Stabilizing Cartridge
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
10-451-115 Stabilizing Cartridge
10-451-038 Pretreatment Cartridge
10-451-021 Filter with Stabilizer
10-451-041 Pretreatment Filter
10-451-222 Filter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 10-451-115 Supplier Thermo Scientific™ Supplier No. 06.5452
Add to Cart
Edge
Add to Cart
Choose one of these Thermo Scientific™ Pretreatment Cartridges and Filters when the quality of your feed water doesn't match the requirements for your point of use water purification system.
Use Thermo Scientific™ Pretreatment Cartridges and Filters when the quality of your feed water doesn't match the requirements for your Thermo Scientific™ Barnstead™ water purification system.
Use pretreatment when your feed water quality doesn't match the requirements for your system.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) LabTower TII, Pacific RO, Pacific TII
Type Stabilizing Cartridge
Includes Hardness Stabilizing Cartridge
Length (English) 10 ft.
Height (English) 10 ft.
Height (Metric) 25.4 cm