Learn More
Healix PREVAL SPRAYER ONLY
Supplier:  Healix PVE267
Encompass_Preferred
Comes complete with power unit and glass bottle Get perfect color matching with this handy little tool Hundreds of uses! No cords, batteries, or hoses to get in the way Preval is the perfect mix-your-own pocket-sized sprayer for every little job where you need quality and fast results It gives you the advantage of perfect color matching every time, because it lets you mix and apply your own paints. Preval can handle all kinds of liquids: paints, lacquers, shellac, cleaners, solvents, gel coats, and much more. Each Preval Spray Gun comes with a modular power unit and refillable container jar with cap. Each power unit sprays up to 16oz. of product. There are no CFC's, so you know it's safe for the environment.
Catalog No. NC9066939
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.