Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ prfA Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144446
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144446 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144446 Supplier Invitrogen™ Supplier No. PA5144446
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

prfA Polyclonal Antibody for Western Blot, ELISA

Positively regulates expression of listeriolysin, of 1-phosphadidylinositol phosphodiesterase (PI-PLC) and other virulence factors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen prfA
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.01 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene prfA
Gene Accession No. P22262
Gene Alias Listeriolysin regulatory protein; prfA
Gene Symbols prfA
Host Species Rabbit
Immunogen Recombinant Listeria monocytogenes serovar 1/2a (strain ATCC BAA-679/EGD-e) Listeriolysin regulatory protein (1-237aa).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 987031
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.