Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRG2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530130
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530130 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530130 Supplier Invitrogen™ Supplier No. PA530130
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Jurkat, K562, THP-1, HL-60. Predicted reactivity: Human (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is the predominant constituent of the crystalline core of the eosinophil granule. High levels of the proform of this protein are also present in placenta and pregnancy serum, where it exists as a complex with several other proteins including pregnancy-associated plasma protein A (PAPPA), angiotensinogen (AGT), and C3dg. This protein may be involved in antiparasitic defense mechanisms as a cytotoxin and helminthotoxin, and in immune hypersensitivity reactions. It is directly implicated in epithelial cell damage, exfoliation, and bronchospasm in allergic diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRG2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.18 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene PRG2
Gene Accession No. P13727
Gene Alias BMPG; bone marrow proteoglycan; bone-marrow proteoglycan; EMBP; Eosinophil granule major basic protein; eosinophil major basic protein; LPR3; major basic protein 1; MBP; MBP1; Mbp-1; mMBP; mMBP-1; natural killer cell activator; Pregnancy-associated major basic protein; PRG2; PRG-2; proMBP; Proteoglycan 2; proteoglycan 2 preproprotein; proteoglycan 2, bone marrow; proteoglycan 2, bone marrow (natural killer cell activator, eosinophil granule major basic protein); proteoglycan 2, pro eosinophil major basic protein
Gene Symbols PRG2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PRG2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5553
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.