Learn More
PRH1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 214601AP150UL
Description
This antibody specifically recognizes the 17 kDa protein.
The PRH1 gene encodes a member of the heterogeneous family of proline-rich salivary glycoproteins. The encoded preproprotein undergoes proteolytic processing to generate one or more mature isoforms before secretion from the parotid and submandibular/sublingual glands. In western population this locus is commonly biallelic and encodes proline-rich protein (PRP) isoforms, PRP-1 and PRP-2. The reference genome encodes the PRP-1 allele. Certain alleles of this gene are associated with susceptibility to dental caries. This gene is located in a cluster of closely related salivary proline-rich proteins on chromosome 12.Specifications
| PRH1 | |
| Polyclonal | |
| Unconjugated | |
| PRH1 | |
| Db F, Db s, Pa, Parotid acidic protein, Parotid double band protein, PIF F, PIF S, Pr1/Pr2, PRH1, PRH1/2, PRH2, Protein A Peptide P C, Protein C, PRP 1/PRP 2, PRP 3/PRP 4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5554 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P02810 | |
| PRH1 | |
| PRH1 Fusion Protein Ag14833 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.