Learn More
Apexbio Technology LLC Rigosertib 592542-59-1 10mg
SDP
Supplier:  Apexbio Technology LLC B128810
Encompass_Preferred
Rigosertib (CAS 592542-59-1) is a small molecule that functions as a dual inhibitor of phosphoinositide 3-kinase (PI3K) and polo-like kinase 1 (PLK1) In cellular models Rigosertib suppresses the PI3K/AKT signaling pathway downregulates cyclin D1 induces pro-apoptotic proteins NOXA and BIM and activates the JNK pathway It promotes apoptosis in a range of human tumor cell lines including breast prostate ovarian pancreatic small cell lung colorectal cancers and melanoma with reported IC50 values typically between 50 90 nM Rigosertib is also active against drug-resistant tumor lines and influences cell cycle progression causing arrest at G1 and G2/M phases in both normal and malignant cells This profile supports its utility in cancer research particularly studies of apoptosis cell cycle regulation and kinase signaling pathways
Catalog No. 50-000-88206
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.