Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prilocaine, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18605815 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-815 25 g Poly Bottle
18-605-814 5 g Amber Glass Bottle
18-605-816 100 g Poly Bottle
18-605-817 500 g Poly Pail
18-605-818 1 kg Poly Pail
18-605-819 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-605-815 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P195425GM
Add to Cart
Add to Cart

P1954, 721-50-6, C16H22N2O3

Spectrum™ Chemical Prilocaine, USP is a local anesthetic used most often for infiltration anesthesia in dentistry. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 721-50-6
Molecular Formula C13H20N2O
Molecular Weight (g/mol) 220.32
MDL Number MFCD00048681
InChI Key MVFGUOIZUNYYSO-UHFFFAOYNA-N
IUPAC Name N-(2-methylphenyl)-2-(propylamino)propanamide
SMILES CCCNC(C)C(=O)NC1=CC=CC=C1C

Specifications

CAS Max % 1
Ignition Residue 0.001
Packaging Poly Bottle
Quantity 25 g
Percent Purity 98 to 102%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.