Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam PRIMA-1MET, P53 Activity, >98% Purity, Solid, 10 mg
Description
- Molecular Weight: 199.25 Da
- Purity: >98%
- Form: Solid
- CAS Number: 5291-32-7
- Key Features: Methylated derivative of PRIMA-1 (ab141641); restores wild-type conformation and function to mutant p53; triggers apoptosis in tumor cells; acts synergistically with chemotherapeutic agents; inhibits thioredoxin reductase 1 (TrxR1).
- Storage: Store at +4°C.
- Shipping: Ambient - Can Ship with Ice.
- Composition: 100% 2-(hydroxymethyl)-2-(methoxymethyl)-1-azabicyclo[2.2.2]octan-3-one.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Content And Storage | Store at 4°C. |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Target Molecule | p53, Thioredoxin Reductase 1 |
| Alternate Part Number | APR-246 |
| Buffer | 100% 2-(hydroxymethyl)-2-(methoxymethyl)-1-azabicyclo[2.2.2]octan-3-one |
| Form | Solid |
| Formulation | 100% 2-(hydroxymethyl)-2-(methoxymethyl)-1-azabicyclo[2.2.2]octan-3-one |
| Quantity | 10 mg |
| For Use With (Application) | p53 activity, apoptosis induction, tumor cell proliferation inhibition, thioredoxin reductase 1 inhibition |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?