Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam PRIMA-1MET, p53 Activity, >98% Purity, Solid, 25 mg

Catalog No. 502704986
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4986 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4986 Supplier Abcam Supplier No. AB14597425MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methylated derivative of PRIMA-1 that restores mutant p53 function and induces apoptosis in tumor cells.

  • Molecular weight: 199.25 Da
  • Purity: >98%
  • Physical form: Solid
  • Function: Methylated derivative of PRIMA-1 that restores wild-type conformation and function to mutant p53 and triggers apoptosis in tumor cells
  • Mechanism of action: Inhibits thioredoxin reductase 1 (TrxR1)
  • Applications: Inhibits tumor cell proliferation and acts synergistically with chemotherapeutic agents
  • Storage: Store at +4°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule p53, Thioredoxin Reductase 1
Shipping Condition Ambient
Form Solid
Alternate Part Number APR-246
Buffer 100% 2-(hydroxymethyl)-2-(methoxymethyl)-1-azabicyclo[2.2.2]octan-3-one
Content And Storage Store at 4°C.
Formulation 100% 2-(hydroxymethyl)-2-(methoxymethyl)-1-azabicyclo[2.2.2]octan-3-one
Quantity 25 mg
For Use With (Application) p53 activity, apoptosis induction, tumor cell proliferation inhibition, thioredoxin reductase 1 inhibition
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.