Learn More
Bio-Serv™ Prima-Treats™ Animal Diet

Description
- Comes in variety of flavors
- Nutritionally complete treat that compliments enrichment programs
- Less labor intensive and more sanitary than perishable fruit
- Excellent source of vitamin C
- Nutritionally assayed
- Weight: 5g (0.18 oz.)
- Shape: Wafer
- Packaging: 250/Jar and 1000/Box.
Specifications
Specifications
| Product Type | Animal Diet |
| Calories | 16.4 |
| Components | Dextrose, Casein, Sucrose, Cellulose, Mineral Mix, Canola Oil, Natural and Artificial Flavors, Magnesium Silicate, Choline Dihydrogen Citrate, Vitamin Mix, Calcium Silicate, Ascorbic Acid, DL-Methionine, Magnesium Stearate, Food Dye, BHA |
| Flavor | Assorted Fruit |
| Particle Shape | Wafer |
| Quantity | 1000/Box |
| Storage Requirements | 12 Months, Cool Dry |
| Diameter (English) | 1 in. |
| Diameter (Metric) | 2.54 cm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.