Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio-Serv™ Prima-Treats™ Animal Diet
SDP

Catalog No. 14726457
Encompass
Change view
Click to view available options
Flavor:
Apple
Assorted Fruit
Banana
Blueberry
Cherry
Grape
Orange
Very Berry
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flavor
14-726-457 Cherry
14-726-454 Apple
14-726-455 Assorted Fruit
14-726-456 Banana
14-726-459 Grape
14-726-462 Very Berry
14-726-460 Orange
14-726-698 Blueberry
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 14-726-457 Supplier Bio-Serv™ Supplier No. F0296
Add to Cart
Add to Cart

Healthy treat used for training or positive reward following a less desirable procedure.

  • Comes in variety of flavors
  • Nutritionally complete treat that compliments enrichment programs
  • Less labor intensive and more sanitary than perishable fruit
  • Excellent source of vitamin C
  • Nutritionally assayed
  • Weight: 5g (0.18 oz.)
  • Shape: Wafer
  • Packaging: 250/Jar and 1000/Box.

Specifications

Product Type Animal Diet
Calories 16.4 kcal/Wafer
Components Dextrose, Casein, Sucrose, Cellulose, Mineral Mix, Canola Oil, Natural and Artificial Flavors, Magnesium Silicate, Choline Dihydrogen Citrate, Vitamin Mix, Calcium Silicate, Ascorbic Acid, DL-Methionine, Magnesium Stearate, Food Dye, BHA
Flavor Cherry
Particle Shape Wafer
Quantity 1000/Box
Storage Requirements 12 Months, Cool Dry
Diameter (English) 1 in.
Diameter (Metric) 2.54 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.