Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ Primair™ Hood Replacements

Catalog No. p-7189511 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
For Use With (Equipment):
Compact Air 200 Series PAPR, CF1000 Series continuous flow airline
PA121
PA131
Quantity:
10/Cs.
3/Cs.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Quantity
19-085-559 PA121 3/Cs.
19-085-560 PA131 3/Cs.
19-085-562 Compact Air 200 Series PAPR, CF1000 Series continuous flow airline 10/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-085-559 Supplier Honeywell™ Supplier No. PA122
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Honeywell Primair™ Hood Replacements are available with under the chin, nylon collar and full bib versions

  • Hoods with collar and full bib have an assigned protection factor (APF) of 1000
  • Manifold directs air over lens to reduce fogging
  • PA121 and PA141 are coated for protection from intermittent splash
  • Primair 100 Series hoods for use with Compact Air 200 Series PAPR and CF1000 Series continuous flow airline feature a comfortable headgear that directs air over the lens for cooling comfort
  • Triple adjustment headgear for a comfortable fit

Specifications

Color White
Product Type Respirator Hood
For Use With (Equipment) PA121
Model PA122
Quantity 3/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.